Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Bag
GREENER_CHOICE
Product Code. p-7203250 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
2 x 2 in.
3 x 1 in.
3 x 2 in.
3 x 2.5 in.
3 x 3 in.
4 x 1 in.
4 x 2 in.
5 x 1 in.
5 x 2 in.
5 x 3 in.
6 x 1 in.
6 x 2 in.
6 x 3 in.
7 x 1 in.
7 x 2 in.
8 x 1 in.
8 x 2 in.
8 x 3 in.
9 x 2 in.
9 x 3 in.
10 x 2 in.
10 x 3 in.
11 x 1 in.
11 x 2 in.
12 x 2 in.
12 x 3 in.
13 x 2 in.
Capacity (English):
3 oz.
4 oz.
5 oz.
6 oz.
7 oz.
7.5 oz.
8 oz.
9 oz.
10 oz.
11 oz.
14 oz.
15 oz.
16 oz.
18 oz.
20 oz.
22 oz.
24 oz.
26 oz.
27 oz.
30 oz.
36 oz.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) Capacity (English) unitSize
17767857 3 x 1 in. 3 oz. Pack of 1000
17777857 4 x 1 in. 4 oz. Pack of 500
17787857 5 x 1 in. 5 oz. Pack of 500
17797857 6 x 1 in. 6 oz. Pack of 500
17707867 7 x 1 in. 7 oz. Pack of 500
17717867 8 x 1 in. 8 oz. Pack of 500
17727867 11 x 1 in. 11 oz. Pack of 500
17737867 3 x 2.5 in. 7.5 oz. Pack of 500
17747867 2 x 2 in. 4 oz. Pack of 500
17757867 3 x 2 in. 6 oz. Pack of 500
17767867 4 x 2 in. 8 oz. Pack of 500
17777867 5 x 2 in. 10 oz. Pack of 500
17797867 6 x 2 in. 10 oz. Pack of 500
17770368 7 x 2 in. 14 oz. Pack of 500
17707877 8 x 2 in. 16 oz. Pack of 500
17717877 9 x 2 in. 18 oz. Pack of 500
17727877 10 x 2 in. 20 oz. Pack of 500
17737877 11 x 2 in. 22 oz. Pack of 500
17757877 12 x 2 in. 24 oz. Pack of 250
17767877 13 x 2 in. 26 oz. Pack of 500
17777877 3 x 3 in. 9 oz. Pack of 500
17787877 5 x 3 in. 15 oz. Pack of 500
17797877 6 x 3 in. 18 oz. Pack of 500
17780368 8 x 3 in. 24 oz. Pack of 500
17707887 9 x 3 in. 27 oz. Pack of 500
17717887 10 x 3 in. 30 oz. Pack of 250
17727887 12 x 3 in. 36 oz. Pack of 500
27 options
Product Code. 17767857
Brand: Keystone Cleanroom Products BGT2A0103D10
Only null left
Add to basket
Add to basket
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. These filling needle bags stay in place via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically removed with forceps after the filling needles are set in place, without having to cut, or remove particulate generating autoclave tape.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Capacity (English) 3 oz.
Capacity (Metric) 88 mL
Certifications/Compliance COC
Closure Type Drawstring
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W) 3 x 1 in.
Length (English) 3 in.
Length (Metric) 7.6 cm
Width (English) 1 in.
Width (Metric) 2.5 cm
Product Type Autoclave Bag