Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave 2X Large Bag
GREENER_CHOICE
Product Code. 17768127 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
28 x 50 in.
34 x 34 in.
36 x 20 in.
36 x 36 in.
36 x 42 in.
36 x 46 in.
38 x 38 in.
40 x 40 in.
44 x 42 in.
46 x 26 in.
48 x 24 in.
48 x 32 in.
48 x 36 in.
48 x 48 in.
48 x 60 in.
50 x 44 in.
50 x 48 in.
50 x 62 in.
60 x 40 in.
60 x 60 in.
64 x 36 in.
64 x 48 in.
70 x 18 in.
72 x 36 in.
Unit Size:
Each
Pack of 100
Pack of 125
Pack of 250
Product Code. Dimensions (L x W) unitSize
17768127 48 x 32 in. Pack of 125
17718067 70 x 18 in. Pack of 250
17788097 48 x 24 in. Pack of 250
17768107 46 x 26 in. Pack of 250
17798127 34 x 34 in. Pack of 250
17852041 36 x 20 in. Pack of 250
17768137 36 x 36 in. Pack of 250
17788137 36 x 42 in. Pack of 125
17708147 36 x 46 in. Pack of 125
17738147 48 x 36 in. Pack of 125
17778147 64 x 36 in. Pack of 125
17798147 72 x 36 in. Pack of 125
17718157 38 x 38 in. Pack of 125
17748157 40 x 40 in. Pack of 125
17778157 60 x 40 in. Pack of 125
17738167 44 x 42 in. Pack of 125
17768167 50 x 44 in. Pack of 125
17710378 48 x 48 in. Pack of 125
17708177 50 x 48 in. Pack of 125
17949519 48 x 60 in. Each
17728177 64 x 48 in. Pack of 100
17748177 28 x 50 in. Pack of 125
17583326 60 x 60 in. Pack of 100
17778177 50 x 62 in. Pack of 125
24 options
Product Code. 17768127
Brand: Keystone Cleanroom Products BGT2A3248D
Only null left
Add to basket
Add to basket
Designed for steam sterilization of large or odd shaped pharmaceutical equipment change parts. Tyvek™ Autoclave Bag is closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Capacity (English) 1488 oz.
Capacity (Metric) 44 L
Certifications/Compliance COC
Closure Type Drawstring
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W) 48 x 32 in.
Length (English) 48 in.
Length (Metric) 122 cm
Width (English) 32 in.
Width (Metric) 81.3 cm
Product Type Autoclave Bag