Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave 2X Large Bag
GREENER_CHOICE
Product Code. 17583326 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
28 x 50 in.
34 x 34 in.
36 x 20 in.
36 x 36 in.
36 x 42 in.
36 x 46 in.
38 x 38 in.
40 x 40 in.
44 x 42 in.
46 x 26 in.
48 x 24 in.
48 x 32 in.
48 x 36 in.
48 x 48 in.
48 x 60 in.
50 x 44 in.
50 x 48 in.
50 x 62 in.
60 x 40 in.
60 x 60 in.
64 x 36 in.
64 x 48 in.
70 x 18 in.
72 x 36 in.
Unit Size:
Each
Pack of 100
Pack of 125
Pack of 250
Product Code. 17583326
Brand: Keystone Cleanroom Products BGT2A6060D
Only null left
Add to basket
Add to basket
Designed for steam sterilization of large or odd shaped pharmaceutical equipment change parts. Tyvek™ Autoclave Bag is closed via the adjustable Tyvek™ drawstring closure system. The bags can be aseptically opened without having to cut or remove the particulate generating autoclave tape.
  • Designed for steam sterilization
  • Tyvek™ on both sides for maximum breathability during sterilization
  • Eliminates need to cut material and the use of autoclave tape
  • Fast and consistent for equipment preparation area
  • Creates secondary level of protection during aseptic presentation of equipment
  • Sealed seams eliminate sewing threads and greatly reduce particulates
  • Resists tearing and punctures
  • Superior microbial barrier
  • Lint-free Category 1 (Helmke Drum)
  • Manufactured and packaged in a ISO 6 cleanroom environment
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Drawstring
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Includes Steam Indicator
Material Tyvek
Sterility Non-sterile
Quantity 100 Pk.
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W) 60 x 60 in.
Length (English) 60 in.
Length (Metric) 152.4 cm
Width (English) 60 in.
Width (Metric) 152.4 cm
Product Type Autoclave Bag