Learn More
Leica Microsystems Type G Immersion Liquid
Product Code. 15336741
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15336741 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15336741 Supplier Leica Microsystems Supplier No. 11513910
Glycerol-formulated immersion liquid (ISO 836). Leica Microsystems™ Type G Immersion Liquid avoids refractive index mismatching encountered with oil immersion fluids and glycerol-based mounting media.
  • Increases objective resolution.
  • Decreases amount of light refraction and reflection from the specimen.
  • Increases the ability of the objective to capture otherwise deviated light.
  • Compatible with glycerol-based mounting media.
TRUSTED_SUSTAINABILITY
Specifications
Type Glycerol Immersion Fluid
For Use With (Application) Glycerol mounting media
Quantity 10 mL