Learn More
Masterflex™ Tygon™ Lab Tubing
Product Code. 15446633
Click to view available options
Diameter (Metric) Inner:
4 mm
4.8 mm
8 mm
9.6 mm
19.1 mm
Diameter (Metric) Outer:
5.6 mm
6.4 mm
12.8 mm
14.4 mm
16 mm
25.4 mm
Maximum Pressure:
14 psi
17 psi
24 psi
25 psi
30 psi
Thickness (Metric):
0.8 mm
1.6 mm
3.2 mm
Unit Size:
15m
50ft
Each
Product Code. 15446633
Brand: Masterflex™ 0640796
Only null left
Add to basket
Add to basket
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon™ Lab Tubing handles virtually all inorganic chemicals found in the lab.
  • Alternative to DEHP tubing
  • New formulation delivers the same excellent performance
  • Contains no BPA
  • Outstanding chemical resistance
  • Lot-to-lot consistency for reproducible results
  • Increases productivity in peristaltic pumps – outlasts other clear tubing 2 to 1
  • Smooth, polished inner bore
  • Non-oxidizing and non-contaminating
  • Slips easily over fittings and grips securely
  • Coils marked at 1-foot intervals for easy measuring
Specifications
Certifications/Compliance FDA compliant; meets USP Class VI and NSF 51 requirements
Autoclavable Autoclavable
Diameter (Metric) Inner 19.1 mm
Material Tygon E-3603
Diameter (Metric) Outer 25.4 mm
Overall Length (Metric) 15.24 m
Thickness (Metric) 3.2 mm
For Use With (Application) General laboratory, food and beverage, biopharmaceutical, analytical instruments
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 1 Pack
Sterility Sterile
Color Transparent
Maximum Pressure 14 psi
Max. Operating Temperature (Metric) 74°C