Learn More
Masterflex™ Tygon™ Lab Tubing
Description
- Alternative to DEHP tubing
- New formulation delivers the same excellent performance
- Contains no BPA
- Outstanding chemical resistance
- Lot-to-lot consistency for reproducible results
- Increases productivity in peristaltic pumps – outlasts other clear tubing 2 to 1
- Smooth, polished inner bore
- Non-oxidizing and non-contaminating
- Slips easily over fittings and grips securely
- Coils marked at 1-foot intervals for easy measuring
Specifications
Specifications
| Certifications/Compliance | FDA compliant; meets USP Class VI and NSF 51 requirements |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 9.6 mm |
| Material | Tygon E-3603 |
| Diameter (Metric) Outer | 16 mm |
| Overall Length (Metric) | 15.24 m |
| Thickness (Metric) | 3.2 mm |
| For Use With (Application) | General laboratory, food and beverage, biopharmaceutical, analytical instruments |
| Min. Operating Temperature (Metric) | -46°C |
| Packaging Quantity | 1 Pack |
| Show More |