Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Description
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Specifications
Specifications
| Color Code | Red/Red |
| Diameter (Metric) Inner | 1.14 mm |
| Material | Tygon F4040 |
| Overall Length (Metric) | 381 mm |
| Thickness (Metric) | 0.85 mm |
| Packaging Quantity | 12 Pack |