Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.64 mm
0.89 mm
1.02 mm
1.14 mm
1.3 mm
1.42 mm
1.5 mm
1.85 mm
2.06 mm
2.29 mm
Unit Size:
Pack of 12
Description
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
Specifications
Specifications
| Color Code | Orange/Orange |
| Diameter (Metric) Inner | 0.89 mm |
| Material | Tygon F4040 |
| Overall Length (Metric) | 381 mm |
| Thickness (Metric) | 0.85 mm |
| Packaging Quantity | 12 Pack |