Learn More
Masterflex™ Tygon™ 2 Stop Hydrocarbon Tubing
Product Code. 11907865
Change view
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.64 mm
0.89 mm
1.02 mm
1.14 mm
1.3 mm
1.42 mm
1.5 mm
1.85 mm
2.06 mm
2.29 mm
Unit Size:
Pack of 12
Product Code. Diameter (Metric) Inner unitSize
11907865 0.25 mm Pack of 12
11937865 0.64 mm Pack of 12
11997865 1.42 mm Pack of 12
11957865 0.89 mm Pack of 12
11967865 1.02 mm Pack of 12
11987865 1.3 mm Pack of 12
11977865 1.14 mm Pack of 12
11907875 1.5 mm Pack of 12
11927875 1.85 mm Pack of 12
11937875 2.06 mm Pack of 12
11947875 2.29 mm Pack of 12
11 options
Product Code. 11907865
Brand: Masterflex™ 9560612
Only null left
Add to basket
Add to basket
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 2 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Color Code Orange/Blue
Diameter (Metric) Inner 0.25 mm
Material Tygon F4040
Overall Length (Metric) 381 mm
Thickness (Metric) 0.90 mm
Packaging Quantity 12 Pack