Learn More
Ismatec™ Tygon™ E-LFL L/S™ 2-Stop Precision Pump Tubing
Click to view available options
Tubing Size:
14
16
Unit Size:
Pack of 8
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Long life - won't age or oxidize
- Fits tightly to glass and metal & bends to small radius curves
- Nontoxic
- Transparent
- For use with Masterflex™ multichannel pump heads
Specifications
Specifications
| Certifications/Compliance | USP; RoHS; Non-Phthalate; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Prop 65 NA (no Prop 65 chemicals) |
| Diameter (Metric) Inner | 1.6 mm |
| Material | Tygon E-LFL |
| Overall Length (English) | 1.25 ft. |
| Overall Length (Metric) | 0.38 m |
| Min. Operating Temperature (Metric) | 0°C |
| Packaging Quantity | Pk. of 8 |
| Tubing Size | 14 |
| Flow Rate | 130 mL/min |
| Hose Barb | 1/16 in. |
| Show More |