Learn More
Saint Gobain Life Sciences Tygon™ 3350 Sanitary Silicone Tubing
Product Code. p-8365011
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
15 mm
18 mm
20 mm
Diameter (Metric) Outer:
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
10 mm
11 mm
12 mm
14 mm
21 mm
24 mm
27 mm
Unit Size:
15m
Each
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer unitSize
12356899 3 mm 6 mm Each
12376899 4 mm 7 mm Each
12396899 5 mm 8 mm Each
12306909 6 mm 9 mm Each
12346909 8 mm 11 mm Each
12386899 4 mm 8 mm Each
12316909 6 mm 10 mm Each
12356909 8 mm 12 mm Each
12376909 10 mm 14 mm Each
12326909 6 mm 12 mm Each
12396909 15 mm 21 mm Each
12306919 18 mm 24 mm Each
12336899 2 mm 4 mm Each
12346899 3 mm 5 mm Each
12326899 1 mm 3 mm Each
12336909 7 mm 10 mm 15m
12316919 20 mm 27 mm Each
17 options
Product Code. 12356899
Brand: Saint Gobain Life Sciences T330D153
Only null left
Add to basket
Add to basket
Designed to reduce the risk of particle entrapment and microscopic build-up during fluid transfer. Tygon™ 3350 Sanitary Silicone Tubing features an ultra-smooth inner surface and reduced surface area for improved flow characteristics.
  • Ultra-smooth inner bore reduces potential for particle entrapment
  • Minimal extractables help maintain fluid integrity
  • Documented biocompatibility for sensitive applications
  • Excellent fluid flow characteristics
  • Fully tested to ISO 10993 standards to facilitate validation process
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NSF-51, 3-A, FDA
Diameter (English) Inner 0.12 in.
Diameter (Metric) Inner 3 mm
Material Silicone
Diameter (English) Outer 0.24 in.
Diameter (Metric) Outer 6 mm
Overall Length (English) 49.2 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.06 in.
Thickness (Metric) 1.5 mm
For Use With (Application) Food and beverage dispensing
Min. Operating Temperature (English) -112°F
Min. Operating Temperature (Metric) -80°C
Color Clear
Hardness 50 (Shore A)
Max. Operating Temperature (English) 399.2°F
Max. Operating Temperature (Metric) 204°C
Product Type Tubing