Learn More
Masterflex™ Tygon 2-stopped Pump Tubing
Product Code. 15327953
Change view
Click to view available options
Diameter (Metric) Inner:
0.89 mm
Unit Size:
Pack of 12
Product Code. Diameter (Metric) Inner unitSize
15327953 0.89 mm Pack of 12
1 options
Product Code. 15327953
Brand: Masterflex™ 9644926
Only null left
Specifications
Diameter (Metric) Inner 0.89 mm
Material Tygon E-LFL
Overall Length (Metric) 38 cm
For Use With (Application) almost any pumping application
Flow Rate 0.011 to 24mL/min.