missing translation for 'onlineSavingsMsg'
Learn More

TY-52156, MedChemExpress

Product Code. p-200100266
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487686

Brand: Medchem Express HY19736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM.

TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM.

Specifications

Chemical Name or Material TY-52156
Physical Form Solid
CAS 934369-14-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H19Cl2N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (274.52 mM)
SMILES ClC1=CC=C(/N=C(NNC2=CC=C(Cl)C=C2)/C(C(C)(C)C)=O)C=C1
Molecular Weight (g/mol) 364.27
Formula Weight 364.27
Percent Purity 99.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.