Learn More
TY-52156, MedChemExpress
Product Code. 16487686
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487686
Brand: Medchem Express HY19736
MedChemExpress TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM.
TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM.
Specifications
Chemical Name or Material TY-52156
Physical Form Solid
CAS 934369-14-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H19Cl2N3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (274.52 mM)
SMILES ClC1=CC=C(/N=C(NNC2=CC=C(Cl)C=C2)/C(C(C)(C)C)=O)C=C1
Molecular Weight (g/mol) 364.27
Formula Weight 364.27
Percent Purity 99.62%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.