Learn More
TXNL6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11203-1-AP-150UL
 View more versions of this product
Product Code. 16878123
  • 4836.10 NOK / 150µL
Description
Retinitis pigmentosa (RP) is a disease that leads to blindness by degeneration of cone photoreceptors. Rods produce factors required for cone viability. The protein encoded by this gene is one of those factors and is similar to a truncated form of thioredoxin. This gene has been proposed to have therapeutic value against RP.
Specifications
Specifications
TXNL6
Polyclonal
Unconjugated
NXNL1
nucleoredoxin like 1, Nucleoredoxin like protein 1, NXNL1, RDCVF, Thioredoxin like protein 6, TXNL6
Rabbit
Antigen Affinity Chromatography
RUO
115861, 234404
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VC33, Q96CM4
Nxnl1
TXNL6 Fusion Protein Ag1691
150 μL
Primary
Human, Mouse