missing translation for 'onlineSavingsMsg'
Learn More

TX1-85-1, MedChemExpress

Product Code. p-200099785
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16495917

Brand: Medchem Express HY100848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. TX1-85-1 is also the first selective Her3 ligand, which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling.

TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. TX1-85-1 is also the first selective Her3 ligand, which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling.

Specifications

Chemical Name or Material TX1-85-1
CAS 1603845-32-4
Color White
Purity Grade Notes Research
Molecular Formula C32H36N8O3
Quantity 5 mg
Solubility Information DMSO : 3.85 mg/mL (6.63 mM; Need ultrasonic)
SMILES CC(N1CCN(C2CCC(N3N=C(C4=CC(NC(C=C)=O)=C(OC5=CC=CC=C5)C=C4)C6=C3N=CN=C6N)CC2)CC1)=O
Molecular Weight (g/mol) 580.68
Formula Weight 580.68
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.