Learn More
Thermo Scientific™ TX-750 4 x 750 mL Swinging Bucket Rotor
Product Code. p-4527973 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
250 mL
4 x 750 mL
750 mL
Not Applicable
Type:
Bucket Lid
Microplate/Flask Carrier
O-ring
Swinging Bucket Rotor
For Use With (Equipment):
ClickSeal Biocontainment Lids
Rectangular Buckets
Round Buckets
Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Unit Size:
2 sets
4 sets
Each
Pack of 4
Product Code. Capacity Type For Use With (Equipment) unitSize
15143169 4 x 750 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F Each
10683755 750 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F 4 sets
10606085 Not Applicable Bucket Lid Round Buckets Pack of 4
11874451 Not Applicable O-ring ClickSeal Biocontainment Lids Pack of 4
10247574 250 mL Swinging Bucket Rotor Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F 4 sets
10389874 Not Applicable Bucket Lid Rectangular Buckets Pack of 4
10718975 4 x 750 mL Microplate/Flask Carrier Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F Pack of 4
11848143 4 x 750 mL Microplate/Flask Carrier Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F 2 sets
8 options
Product Code. 15143169
Brand: Thermo Scientific™ 75003180
Increase lab productivity with the 750 mL swinging bucket rotor, delivering maximum flexibility at 4700 rpm.
Increase lab productivity with the Thermo Scientific™ TX-750 4 x 750mL Swinging Bucket Rotor, delivering maximum flexibility at 4700rpm.
Ordering Information:
Buckets, lids and microplate carriers are sold separately; order Rotor Body (75-003-607) and components to complete assembly. For replacement O-rings for lids (set of four) order 75-003-610. Tube adapters are also available separately. Contact your sales representative for information.
Includes:
Rotor manual
Compatible with:
Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
TRUSTED_SUSTAINABILITY
Specifications
Description TX-750 (Rotor body only) w/Slide Coat anti-friction finish
Type Swinging Bucket Rotor
Capacity 4 x 750 mL
Max. RCF 4,816 x g (with 75-003-608 round buckets, RCF varies with bucket type)
Max. Speed 4,700 rpm (with 75-003-608 round buckets, speed varies with bucket type)
Radius (Metric) 195 mm
Max. Tube Length 139 mm
Max. Tube Diameter 98 mm
For Use With (Equipment) Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40, SL 40/40F
Angle 90°
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Biocontainment Yes