Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
- Maximum tube capacity: 4 × 100mL
- Maximum O.D. × L: 48 × 122mm
Specifications
Specifications
| Type | Rotor Tube Adapter |
| Capacity | 1.5/2.0 mL Conical Microtube |
| Max. Tube Diameter | 11 mm |
| Max. Tube Length | 45 mm |