Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Product Code. 10267044 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection or 15 mL Corex /Kimble Tube
100 mL Round-bottom Tube
12 x 124 mm Tube
14 mL Conical Urine Tube
15 mL Blood Collection Tube
15 mL Conical Tube
20 mL Round-bottom Tube
25 mL DIN Round-/Flat-bottom Tube
25 mL Universal Tube
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube
50 mL Conical Tube
50 mL DIN Round-bottom Tube
50 mL Round-bottom Tube
Unit Size:
Pack of 4
Product Code. Capacity unitSize
10267044 12 x 124 mm Tube Pack of 4
10194494 15 mL Conical Tube Pack of 4
10596214 5/7 mL Blood Collection Tube Pack of 4
10596404 100 mL Round-bottom Tube Pack of 4
10338204 50 mL DIN Round-bottom Tube Pack of 4
10082534 50 mL Conical Tube Pack of 4
10216854 25 mL Universal Tube Pack of 4
10297374 25 mL DIN Round-/Flat-bottom Tube Pack of 4
10459834 20 mL Round-bottom Tube Pack of 4
10124364 14 mL Conical Urine Tube Pack of 4
10318064 10 mL Blood Collection or 15 mL Corex /Kimble Tube Pack of 4
10799305 15 mL Blood Collection Tube Pack of 4
10677565 5/7 mL Round-bottom Tube Pack of 4
10431565 1.5/2.0 mL Conical Microtube Pack of 4
10738545 50 mL Round-bottom Tube Pack of 4
15 options
Product Code. 10267044
Brand: Thermo Scientific™ 75003811
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Capacity 12 x 124 mm Tube
Max. Tube Diameter 12 mm
Max. Tube Length 124 mm