Learn More
Köttermann Two Door Tall Storage Cabinet
Click to view available options
Door Type:
Hinged
Sliding
No. of Shelves:
4
None
Dimensions (D x W x H):
350 x 900 x 1920 mm
366 x 900 x 1920 mm
500 x 900 x 1920 mm
516 x 900 x 1920 mm
350 x 1200 x 1920 mm
500 x 1200 x 1920 mm
770 x 1100 x 2250 mm
Unit Size:
Each
Description
- Tall, slender design expands storage with a small footprint
- Two doors protect stored items
- Sliding and hinged door options available
Specifications
Specifications
| Color | White |
| Adjustable | No |
| Cabinet Type | Tall Storage |
| Dimensions (D x W x H) | 366 x 900 x 1920 mm |
| Door Type | Hinged |
| Finish | Epoxy Coated |
| For Use With (Application) | Storage |
| No. of Doors | 2 |
| No. of Drawers | None |
| No. of Shelves | 4 |
| Show More |