missing translation for 'onlineSavingsMsg'
Learn More

TWIST1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16844045 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16844045 20 μL 20µL
16834045 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16844045 Supplier Proteintech Supplier No. 254651AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody reacts specifically with TWIST1 protein.

TWIST1 acts as a transcriptional regulator. It inhibits myogensis by sequestering E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses proinflammatory cytokines such as TNFA and IL1B. TWIST also regulates cranial suture patterning and fusion and gene expression differentially depending on dimer composition. Mutations in the gene are implicated in Saethre-Chotzen syndrome, a disease characterized by coronal synostosis, brachycephaly, low front hairline, facial asymmetry, hypertelorism, broad halluces, and clinodactyly.

Specifications

Antigen TWIST1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TWIST1
Gene Accession No. P26687, Q15672
Gene Alias ACS3, bHLHa38, BPES2, BPES3, H twist, SCS, TWIST, twist homolog 1 (Drosophila), Twist related protein 1, TWIST1
Gene Symbols TWIST1
Host Species Rabbit
Immunogen TWIST1-specific Fusion Protein Ag21924
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22160, 7291
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.