Learn More
LABPLAS Twirl'Em Writing
Product Code. 17930145
Change view
Click to view available options
Dimensions (L x W):
89 x 140 mm
89 x 178 mm
114 x 191 mm
114 x 229 mm
114 x 305 mm
114 x 381 mm
140 x 229 mm
140 x 305 mm
140 x 381 mm
178 x 305 mm
178 x 381 mm
Thickness (Metric):
63.5 μm
76.2 μm
101.6 μm
Unit Size:
Pack of 1000
Product Code. Dimensions (L x W) Thickness (Metric) unitSize
17930145 140 x 381 mm 76.2 μm Pack of 1000
17970275 89 x 140 mm 76.2 μm Pack of 1000
17990105 89 x 178 mm 76.2 μm Pack of 1000
17960135 114 x 305 mm 63.5 μm Pack of 1000
17960275 114 x 381 mm 63.5 μm Pack of 1000
17940155 140 x 229 mm 101.6 μm Pack of 1000
17900225 114 x 191 mm 63.5 μm Pack of 1000
17960295 114 x 229 mm 63.5 μm Pack of 1000
17920115 178 x 305 mm 101.6 μm Pack of 1000
17910225 140 x 305 mm 76.2 μm Pack of 1000
17920185 140 x 229 mm 76.2 μm Pack of 1000
17920155 178 x 305 mm 76.2 μm Pack of 1000
17920205 178 x 381 mm 76.2 μm Pack of 1000
13 options
Product Code. 17930145
Brand: LABPLAS EPR5515
Only null left
Add to basket
Add to basket
Sterile sampling bag with writing area for easy labeling.
  • Twirl'Em Writing
  • Safety tabs
  • Clear, with writing area for easy labeling
  • Closure with 1 round wire and 1 flat wire
  • Available in various sizes
Specifications
Autoclavable Not Autoclavable
Color Clear with White Block
Disposable Single-use
Sterility Sterile
Thickness (Metric) 76.2 μm
Latex Free Yes
Thickness (English) 3 mil.
Dimensions (L x W) 140 x 381 mm
Length (English) 5.5 in.
Length (Metric) 140 mm
Width (English) 15 in.
Width (Metric) 381 mm
Product Type Sample Bag