Learn More
LABPLAS Twirl'Em Sample Bags
Product Code. 17950215 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
178 x 305 mm
254 x 381 mm
Unit Size:
Pack of 1000
Product Code. 17950215
Only null left
Add to basket
Add to basket
Twirl'em sample bags are safe, flexible, contaminant-free and easy to use.
  • Twirl'Em sampling bags
  • Safety tabs
  • Clear, with writing area for easy labeling
  • Closure with 1 round wire and 1 flat wire
  • Available in various sizes
Specifications
Autoclavable Not Autoclavable
Color Clear with White Block
Disposable Single-use
Sterility Sterile
Thickness (Metric) 101.6 μm
Latex Free Yes
Thickness (English) 4 mil.
Dimensions (L x W) 254 x 381 mm
Length (English) 10 in.
Length (Metric) 254 mm
Width (English) 15 in.
Width (Metric) 381 mm
Product Type Sample Bag