missing translation for 'onlineSavingsMsg'
Learn More

TUSC4 Antibody [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30499952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499952 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499952 Supplier Novus Biologicals Supplier No. NBP335395G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUSC4 Polyclonal antibody specifically detects TUSC4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC4
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias G21 protein, Gene 21 protein, homologous to yeast nitrogen permease (candidate tumor suppressor), nitrogen permease regulator 2-like protein, nitrogen permease regulator-like 2 (S. cerevisiae), NPR2, NPR2L2810446G01Rik, NPRL2, Tumor suppressor candidate 4NPR2-like protein, TUSC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human TUSC4 (NP_006536.3).,, Sequence:, MGSGCRIECIFFSEFHPTLGPKITYQVPEDFISRELFDTVQVYIITKPELQNKLITVTAMEKKLIGCPVCIEHKKYSRNALLFNLGFVCDAQAKTCALEPIVKKLAGYLTTLELESSFVSMEESKQKLVPIMTILLEELNASGRCTLPIDESNTIHLKVIEQRPDPPVAQEYDVPVFTKDKEDFFNSQWDLTTQQILPYIDGFRHIQKISAEADVELNLV
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10641
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.