Learn More
Masterflex™ Turbo Prop Open Channel Flowmeter
Product Code. 11700099 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate Range:
0.3 to 15 ft./sec.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Range Unit Size
11700099 0.3 to 15 ft./sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11700099 Supplier Masterflex™ Supplier No. 3292201
  • ONLINE EXCLUSIVE PRICE
    28945.00 NOK
    24733.51 NOK / Pack of 1
    Save 4211.49 NOK
    14% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Specifications
Type Paddle Wheel Flowmeter
Accuracy 0.1 ft./sec.
Flow Rate Range 0.3 to 15 ft./sec.
For Use With (Application) Accurately measure water velocity in open channels and partially filled pipes
Power Supply Details Internal lithium Battery
Temperature (Metric) 70°C