missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPP Techno Plastic Products AG Turbo-Fix Consumable Replacment Filter
Description
- Turbo-Fix
- Pipette aid
- Ideal for controled sensitive pipetting speed
- Lightweight but still strong
Specifications
Specifications
| Description | Replacement filter 0.22 μm for Turbo-Fix nose piece Stabifix |
| Color | Red |
| For Use With | Turbo-Fix Nose Piece Stabifix |
| Material | Various |
| Pore Size | 0.22 μm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?