missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPP Techno Plastic Products AG Turbo-Fix Consumable Replacement Housing
Click to view available options
:
Turbo-fix, pipette aid, yellow
Description
- Turbo-Fix
- Pipette aid
- Ideal for controled sensitive pipetting speed
- Lightweight but still strong
Specifications
Specifications
| Description | Housing for Turbo-Fix |
| Color | Yellow |
| For Use With | Turbo-Fix Pipette Aid |
| Material | Polyamide |
| Model | Turbo-Fix |
| Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction