Learn More
WTW™ Turbidity Standard
Product Code. 10134273
Click to view available options
Turbidity:
0.02, 10, 100,1750 NTU
10,000 NTU
Unit Size:
Each
Product Code. 10134273
Brand: WTW™ 600544
Only null left
Add to basket
Add to basket
Turbidity standards AMCO Clear™ with the highest precision and long-term stability are N.I.S.T. traceable, lot certified and not hazardous to your health.
  • Long-term precision and stability
  • No health hazards
  • Easy to dispose of
  • Adapted to the respective device optics in their particle composition
  • Suitable for applications in the lower measuring range such as drinking water
TRUSTED_SUSTAINABILITY
Specifications
Type Turbidity Standard
For Use With (Equipment) Turb 555 IR
Description Calibration standards
Includes N/A
Turbidity 10,000 NTU