Learn More
WTW™ Turbidimeter Cuvette
Click to view available options
For Use With (Equipment):
Photoflex Series and Turb 430 IR/T Turbidimeter and Photometer
Turb 350/355 IR and 355 T Portable Turbidimeter
Turb 550, Turb 550 IR, Turb 555 and Turb 555 (IR) Laboratory Turbidimeter
Unit Size:
Pack of 3
Beskrivelse
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
Tekniske data
Tekniske data
| Type | Cuvette |
| For Use With (Equipment) | Photoflex Series and Turb 430 IR/T Turbidimeter and Photometer |
| Quantity | 3 Pack |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion