Learn More
Tungstophosphoric Acid Hydrate, Supelco™
Specifications
Specifications
| CAS | 12501-23-4 |
| Melting Point | 107°C |
| Quantity | 100 g |
| Molecular Formula | H3[P(W3O10)4] · X H2O |
| MDL Number | MFCD00149915 |
| Synonym | Tungstophosphoric acid hydrate, Phosphotungstic acid, Tungstophosphoric acid hydrate |
| Molecular Weight (g/mol) | 2880.05 (Anhydrous Basis) |
| Chemical Name or Material | Tungstophosphoric Acid Hydrate |