Learn More
MedChemExpress Tuftsin
Specifications
Specifications
| Purity | 98.0% |
| CAS | 9063-57-4 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 500.59 g/mol |
| Product Type | Natural Products |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (199.76 mM∣Need ultrasonic) |