Learn More
Honeywell™ TUFFSHIELD™ Deep Blue Right Hand Only Glove
Description
- Knitted glove, HPPE, composite fiber and polyamide
- Fully dipped light blue nitrile foam coating
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- The composite fibres have been protected so that they do not come directly in contact with the skin
- The hand of the glove is written on the marking (top of the hand) for an easier recognition of the right glove
- The mix of HPPE and composite fiber to obtain excellent protection levels against cuts
- Certified for contact with food and blue in color to meet the expectations of workers from the food sector
- The HPPE fibre resists cuts, abrasion and detergents
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical Hazards |
| Description | 13 gauge |
| Color | Light Blue |
| Hand | Right |
| Material | Nitrile |
| Size | 8 |
| Length (Metric) | 27 cm |