Learn More
Honeywell™ TUFFSHIELD™ Deep Blue Right Hand Only Glove
Product Code. 15856835
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
Each
Product Code. Size unitSize
15856835 7 Each
15866835 8 Each
15876835 9 Each
15886835 10 Each
15896835 11 Each
5 options
Product Code. 15856835
Brand: Honeywell™ 231292907
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ TUFFSHIELD™ Deep Blue Right Hand Only Glove offers protection against extreme cut risk environments like slaughtering, food, fish, and in the paper industry.
  • Knitted glove, HPPE, composite fiber and polyamide
  • Fully dipped light blue nitrile foam coating
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The composite fibres have been protected so that they do not come directly in contact with the skin
  • The hand of the glove is written on the marking (top of the hand) for an easier recognition of the right glove
  • The mix of HPPE and composite fiber to obtain excellent protection levels against cuts
  • Certified for contact with food and blue in color to meet the expectations of workers from the food sector
  • The HPPE fibre resists cuts, abrasion and detergents
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical Hazards
Description 13 gauge
Color Light Blue
Hand Right
Material Nitrile
Size 7
Length (Metric) 27 cm