missing translation for 'onlineSavingsMsg'
Learn More

Tubuloside A, MedChemExpress

Product Code. p-200099065
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16449268 10 mg 10mg
16431688 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16449268

Brand: Medchem Express HYN2155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tubuloside A is a phenylethanoid glycoside with antioxidative effect and hepatoprotective activity.

Tubuloside A is a phenylethanoid glycoside with antioxidative effect and hepatoprotective activity.

Specifications

Chemical Name or Material Tubuloside A
CAS 112516-05-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C37H48O21
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (60.33 mM; Need ultrasonic)
SMILES OC1C(OC(C)C(O)C1O)OC(C(C(OCCC2=CC(O)=C(O)C=C2)O3)OC(C)=O)C(C3COC(C(C(O)C4O)O)OC4CO)OC(/C=C/C5=CC(O)=C(O)C=C5)=O
Molecular Weight (g/mol) 828.76
Formula Weight 828.76
Percent Purity 99.7%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.