Learn More
Upchurch Scientific Tubing SST 1/8 inch O.D., 0.080 inch (2.03mm)I.D. 5m
Brand:  Upchurch Scientific 02011-76
Tubing SST 1/8 inch O.D., 0.080 inch (2.03mm)I.D. 5m
Product Code. 12962019
  • ONLINE EXCLUSIVE PRICE
    10395.00 NOK
    10009.76 NOK / Each
    Save 385.24 NOK
    3% Off
Only null left
Estimated Shipment: 23-01-2026