Learn More
Hirschmann Tubing Pharmed BPT, ID 6.4mm, roll of 7.5M
Brand:  Hirschmann 95213016
Tubing Pharmed BPT, ID 6.4mm, roll of 7.5M
Product Code. 17720898
  • ONLINE EXCLUSIVE PRICE
    4137.00 NOK
    2690.72 NOK / 7.50m
    Save 1446.28 NOK
    34% Off
Only null left
Estimated Shipment: 02-03-2026