Learn More
Lenz Laborglasintrumente Tubing Connectors, Straight
Product Code. 11903351
Change view
Click to view available options
Shape:
Straight
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shape Unit Size
11903351 Straight Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11903351 Supplier Lenz Laborglasintrumente Supplier No. 01600011
  • ONLINE EXCLUSIVE PRICE
    147.00 NOK
    128.57 NOK / Pack of 10
    Save 18.43 NOK
    12% Off
Estimated Shipment: 02-12-2026
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Tubing connectors
  • Straight
  • With two glass olives
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Diameter (Metric) Outer 11 mm
Shape Straight