Learn More
Lauda™ Tubing Connector For LAUDA Thermostats and Chillers
Brand:  Lauda™ EOA 032
Additional Details : Weight : 0.01000kg
Product Code. 12062412
  • 88.80 NOK / Each
Estimated Shipment: 17-06-2024
Description
Order Info
dimFitsTubing
Specifications
Specifications
For 8 to 11 to 4 to 7 mm Polymer Tubings
Tubing Connector
1 Ea.
100°C