Learn More
TUBG2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28009-1-AP-20UL
 View more versions of this product
Product Code. 17231396
  • 1710.33 NOK / 20µL
Description
The TUBG2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TUBG2. This antibody recognizes human, mouse, rat antigen. The TUBG2 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
TUBG2
Polyclonal
Unconjugated
20 μL
27175
Liquid
Western Blot, ELISA
750 μg/mL
Rabbit
Primary
Human, Mouse, Rat