Description
- For use with all GX and GE model transilluminators
- 254nm wavelength
- Small wave
Specifications
Specifications
| Intensity | Fixed |
| Wavelength | 254nm |
| Description | Short wave UV tube |
| For Use With (Equipment) | All GX/GE model transilluminators |
| Type | UV Tube Short Wave |