Learn More
Thermo Scientific™ Tube Racks for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers
Product Code. 17801895 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
24 Tubes
40 Tubes
72 Tubes
Dimensions (L x W x H):
240 x 118 x 83 mm
276 x 118 x 109 mm
307 x 132 x 109 mm
330 x 132 x 109 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) unitSize
17801895 72 Tubes 240 x 118 x 83 mm Each
17851875 40 Tubes 276 x 118 x 109 mm Each
17891885 40 Tubes 330 x 132 x 109 mm Each
17861875 24 Tubes 307 x 132 x 109 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17801895 Supplier Thermo Scientific™ Supplier No. 236090
Mix and match Thermo Scientific™ shaker accessories to suit individual needs in the laboratory. Adjustable Test Tube racks are compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers.
Mix and match Thermo Scientific™ Solaris shaker accessories to suit individual needs in the laboratory. Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers
Mix and match Thermo Scientific™ Solaris™ shaker accessories to suit individual needs in the laboratory.  Compatible with Solaris Open Air, Solaris Temperature Controlled Benchtop Shakers and MaxQ6000 Shakers. The adjustable rack holder allows for the test tube rack to be securely shaken, even when positioned at an angle.
TRUSTED_SUSTAINABILITY
Specifications
Holds 72 Tubes
Tube Size 10 to 13 mm
Product Type Test Tube Rack
Dimensions (L x W x H) 240 x 118 x 83 mm
Description Adjustable tube rack for 10 to 13 mm tubes
For Use With (Equipment) Solaris Open Air Shakers, Solaris Temperature Controlled Shakers, MaxQ 6000 Shakers