Description
- Manufactured from polypropylene
- Seal of EPDM
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs |
| Diameter (Metric) | 13 mm nozzle |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Packaging Quantity | 20/Scale |
| Thread Size | 1/2 in. |
| Width (Metric) | 9 mm |
| Includes | Cap nut |
| Material (Seal) | EPDM |
| Product Type | Tube Nozzle |