Learn More
BOLA Tube Fittings
Product Code. 10268590
Change view
Click to view available options
Diameter (Metric):
4 mm
6 mm
12 mm
14 mm
16 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10268590 4 mm Each
10349371 6 mm Each
10257591 12 mm Each
10718062 14 mm Each
10525201 16 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10268590 Supplier BOLA Supplier No. D50302
  • ONLINE EXCLUSIVE PRICE
    242.55 NOK
    219.99 NOK / Pack of 1
    Save 22.56 NOK
    9% Off
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Tube Fittings PTFE, nut made of glass-fibre reinforced PTFE
  • Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE.
  • Universal chemical resistance, the product is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Diameter (Metric) 4 mm
Autoclavable Autoclavable
Overall Length (Metric) 6 mm
Color White
Flexibility Rigid