Learn More
Ismatec™ Tube Connector, Straight, Plastic
Product Code. 12018528
Change view
Click to view available options
Diameter (Metric) Inner:
For tubing with I.D. 3.0 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12018528 For tubing with I.D. 3.0 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12018528 Supplier Ismatec™ Supplier No. ISM558A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Standard tube connector for pump tubing.
  • Standard connector, straight
  • Suitable for use with pump tubing
  • Acetal resin (POM, Polyoxymethylene) construction
Specifications
Diameter (Metric) Inner For tubing with I.D. 3.0 mm
For Use With Pump tubing
Packaging Quantity 10/Pk
Shape Straight