Learn More
Gosselin™ Tube Closures
Product Code. 11825853
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
Product Type:
Ribbed Closure Plug
Straight Plug
Winged Cap
For Use With (Equipment):
10 mm Tube
11 mm Tube
13 mm Tube
13.5 mm Tube
18.5 mm Tube
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Pack of 2500
Pack of 3000
Pack of 4000
Quantity:
1000/Bag
1500 Bag
2000 Bag
2500 Bag
3000 Bag
4000 Bag
Product Code. 11825853
Brand: Gosselin™ BA11021
Only null left
Add to basket
Add to basket
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ribbed Closure Plug
Material LDPE
Color Blue
Temperature Range (Metric) 80°C to -100°C
For Use With (Equipment) 13 mm Tube