Learn More
Gosselin™ Tube Closures
Product Code. 11778593
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
Product Type:
Ribbed Closure Plug
Straight Plug
Winged Cap
For Use With (Equipment):
10 mm Tube
11 mm Tube
13 mm Tube
13.5 mm Tube
18.5 mm Tube
Quantity:
1000/Bag
1500 Bag
2000 Bag
2500 Bag
3000 Bag
4000 Bag
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Pack of 2500
Pack of 3000
Pack of 4000
Product Code. Color Product Type For Use With (Equipment) Quantity unitSize
11778593 White Straight Plug 11 mm Tube 4000 Bag Pack of 4000
11592762 White Ribbed Closure Plug 11 mm Tube - Pack of 2500
11758593 White Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11748603 Yellow Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11778613 Red Straight Plug 18.5 mm Tube 1000/Bag Pack of 1000
11748593 White Ribbed Closure Plug 10 mm Tube - Pack of 3000
11798613 Green Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11768613 Red Straight Plug 13.5 mm Tube 2000 Bag Pack of 2000
11723313 Natural Winged Cap 13.5 mm Tube 1500 Bag Pack of 1500
11708623 Green Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11798603 Red Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11728603 Blue Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11708613 Red Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11718613 Red Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11738603 Yellow Ribbed Closure Plug 10 mm Tube 3000 Bag Pack of 3000
11758603 Natural Ribbed Closure Plug 11 mm Tube 2500 Bag Pack of 2500
11502772 White Straight Plug 13.5 mm Tube - Pack of 2000
12113791 Blue Ribbed Closure Plug 13.5 mm Tube 1500 Bag Pack of 1500
11825853 Blue Ribbed Closure Plug 13 mm Tube - Pack of 2500
19 options
Product Code. 11778593
Brand: Gosselin™ B11003
Only null left
Add to basket
Add to basket
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
TRUSTED_SUSTAINABILITY
Specifications
Product Type Straight Plug
Material LDPE
Color White
Quantity 4000 Bag
For Use With (Equipment) 11 mm Tube