Learn More
- Home
- Products
- Tubes
- Tube Caps and Closures
- Gosselin™ Tube Closures
Gosselin™ Tube Closures
Product Code. 11728603
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
White
Yellow
Product Type:
Ribbed Closure Plug
Straight Plug
Winged Cap
For Use With (Equipment):
10 mm Tube
11 mm Tube
13 mm Tube
13.5 mm Tube
18.5 mm Tube
Quantity:
1000/Bag
1500 Bag
2000 Bag
2500 Bag
3000 Bag
4000 Bag
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Pack of 2500
Pack of 3000
Pack of 4000
19 product options available for selection
| Product Code. | Color | Product Type | For Use With (Equipment) | Quantity | Unit Size |
|---|---|---|---|---|---|
| 11728603 | Blue | Ribbed Closure Plug | 10 mm Tube | 3000 Bag | Pack of 3000 |
| 11592762 | White | Ribbed Closure Plug | 11 mm Tube | - | Pack of 2500 |
| 11758593 | White | Ribbed Closure Plug | 13.5 mm Tube | 1500 Bag | Pack of 1500 |
| 11748603 | Yellow | Ribbed Closure Plug | 11 mm Tube | 2500 Bag | Pack of 2500 |
| 11778613 | Red | Straight Plug | 18.5 mm Tube | 1000/Bag | Pack of 1000 |
| 11748593 | White | Ribbed Closure Plug | 10 mm Tube | - | Pack of 3000 |
| 11798613 | Green | Ribbed Closure Plug | 10 mm Tube | 3000 Bag | Pack of 3000 |
| 11768613 | Red | Straight Plug | 13.5 mm Tube | 2000 Bag | Pack of 2000 |
| 11723313 | Natural | Winged Cap | 13.5 mm Tube | 1500 Bag | Pack of 1500 |
| 11708623 | Green | Ribbed Closure Plug | 11 mm Tube | 2500 Bag | Pack of 2500 |
| 11798603 | Red | Ribbed Closure Plug | 10 mm Tube | 3000 Bag | Pack of 3000 |
| 11708613 | Red | Ribbed Closure Plug | 11 mm Tube | 2500 Bag | Pack of 2500 |
| 11718613 | Red | Ribbed Closure Plug | 13.5 mm Tube | 1500 Bag | Pack of 1500 |
| 11738603 | Yellow | Ribbed Closure Plug | 10 mm Tube | 3000 Bag | Pack of 3000 |
| 11758603 | Natural | Ribbed Closure Plug | 11 mm Tube | 2500 Bag | Pack of 2500 |
| 11778593 | White | Straight Plug | 11 mm Tube | 4000 Bag | Pack of 4000 |
| 11502772 | White | Straight Plug | 13.5 mm Tube | - | Pack of 2000 |
| 12113791 | Blue | Ribbed Closure Plug | 13.5 mm Tube | 1500 Bag | Pack of 1500 |
| 11825853 | Blue | Ribbed Closure Plug | 13 mm Tube | - | Pack of 2500 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
Denna artikel kan inte returneras.
Se returpolicy
Produktkod.
11728603
Leverantör
Gosselin™
Leverantörsnummer
BA10031
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- All closures vertically ribbed for easier grip
- Protected with a polyethylene outer bag to prevent particle contamination
Specifikationer
Specifikationer
| Product Type | Ribbed Closure Plug |
| Material | LDPE |
| Color | Blue |
| Quantity | 3000 Bag |
| Sterility | Sterile |
| For Use With (Equipment) | 10 mm Tube |
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?