Learn More
JULABO™ Tube Clamps for Reinforced Tubing
JULABO Tube Clamps for Reinforced Tubing are for use with 1.5 in. inner diameter (ID) reinforced tubing.
Brand:  JULABO™ 8970487
Additional Details : Weight : 0.05000kg
Product Code. 17118518
  • 312.00 NOK / Each
Estimated Shipment: 13-06-2024
Description
  • Clamping range: 40 to 60 mm
Specifications
Specifications
Clamping range: 40 to 60 mm
Reinforced Tubing 1.5 in. ID
2 Pk.
6