Learn More
Julabo™ Tube Clamps for Baths
Product Code. 11990318
Change view
Click to view available options
Holds:
0.75 in. I.D. Reinforced tubing
1 in. I.D. Reinforced tubing
12mm /0.5 in. ID. Reinf. tubing
8mm I.D.CR tubing
CR 10 mm ID, Reinf. 8 mm ID tubing
Unit Size:
Pack of 2
Product Code. Holds unitSize
11990318 1 in. I.D. Reinforced tubing Pack of 2
11960318 0.75 in. I.D. Reinforced tubing Pack of 2
11940318 12mm /0.5 in. ID. Reinf. tubing Pack of 2
11990248 CR 10 mm ID, Reinf. 8 mm ID tubing Pack of 2
11980248 8mm I.D.CR tubing Pack of 2
5 options
Product Code. 11990318
Brand: Julabo™ 8970484
Only null left
Specifications
For Use With (Equipment) CF31, CF41 Cryo-compact circulator, F25-HL, F32-HE, F32-HL refrigerated/heating circulator
Type Tube Clamp
Holds 1 in. I.D. Reinforced tubing
Size 5