Learn More
Abnova™ TTLL3 Recombinant Protein
Product Code. 16128122
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16128122
Brand: Abnova™ H00026140P01.10ug
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Accession Number AAH09479
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 26140
Molecular Weight (g/mol) 36.85
Name TTLL3 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen TEGDRNIWIVKPGAKSRGRGIMCMDHLEEMLKLVNGNPVVMKDGKWVVQKYIERPLLIFGTKFDLRQWFLVTDWNPLTVWFYRDSYIRFSTQPFSLKNLDK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp434B103/DKFZp686D076/FLJ13898/HOTTL/MGC120529/MGC120530/MGC120532
Common Name TTLL3
Gene Symbol TTLL3
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution